Titanium powder, hydrided, -325 mesh, 99.99% (metals basis)
Titanium powder, hydrided, -325 mesh, 99.99% (metals basis)
Titanium powder, hydrided, -325 mesh, 99.99% (metals basis)
Thermo Scientific Chemicals

Titanium powder, hydrided, -325 mesh, 99.99% (metals basis)

CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036678.14
also known as 036678-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey
FormPowder
Total Metal Impurities≤0.01% (≤100ppm)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only