Copper shot, 13mm (0.5in) dia, 99.99% (metals basis), oxygen free
Copper shot, 13mm (0.5in) dia, 99.99% (metals basis), oxygen free
Copper shot, 13mm (0.5in) dia, 99.99% (metals basis), oxygen free
Thermo Scientific Chemicals

Copper shot, 13mm (0.5in) dia, 99.99% (metals basis), oxygen free

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 036686.36
also known as 036686-36
Price (USD)
94.65
Online Exclusive
105.00
Save 10.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
94.65
Online Exclusive
105.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max., oxygen free
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only