Hydrobromic acid, ACS, 47.0-49.0%
Hydrobromic acid, ACS, 47.0-49.0%
Hydrobromic acid, ACS, 47.0-49.0%
Hydrobromic acid, ACS, 47.0-49.0%
Thermo Scientific Chemicals

Hydrobromic acid, ACS, 47.0-49.0%

Air Sensitive | CAS: 10035-10-6 | BrH | 80.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
4 x 1 L
Catalog number 036694.M1
also known as 036694-M1
Price (USD)
538.00
Each
Add to cart
Quantity:
4 x 1 L
Request bulk or custom format
Price (USD)
538.00
Each
Add to cart
Chemical Identifiers
CAS10035-10-6
IUPAC Namehydrogen bromide
Molecular FormulaBrH
InChI KeyCPELXLSAUQHCOX-UHFFFAOYSA-N
SMILESBr
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentSe 0.01ppm
Residue on ignition0.002%
Impurity contentPO₄ 0.001%
Impurity contentSulfate and sulfite (as SO₄) 0.003%
Impurity contentCl 0.05%
View more
Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes, and medicines. It is also used in the production of bromine compounds. It is used as an important agent in the industrial processes of isomerization, polymerization, hydration, and dehydration, and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only