Starch, soluble, ACS (for iodometry)
Starch, soluble, ACS (for iodometry)
Starch, soluble, ACS (for iodometry)
Thermo Scientific Chemicals

Starch, soluble, ACS (for iodometry)

For iodometry | CAS: 9005-84-9 | C12H22O11 | 342.297 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 036703.18
also known as 036703-18
Price (USD)
37.00
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
37.00
Each
Chemical Identifiers
CAS9005-84-9
IUPAC Name(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyGUBGYTABKSRVRQ-ASMJPISFSA-N
SMILESOC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Appearance (Color)White
CommentSensitivity: Passes test
Residue on ignition≤0.4%
pH5.0-7.0 (2% solution at 25°C)
View more
In food industry, starch is used as a food preservative. Starch has extensive industrial use as an adhesive or bonding agent. It is also used in paper industry. Starch is used in the procedure to manufacture gypsum wallboards. Starch used in the laundry, also known as clothing starch. Glucose derived from starch may be fermented further to make ethanol or biofuel. Starch is also employed in the petroleum exploration industry. It is interesting to note that starch may be used to make hydrogen by making usage of enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only