3-Methyl-1-butanol, ACS, 98.5+%
3-Methyl-1-butanol, ACS, 98.5+%
3-Methyl-1-butanol, ACS, 98.5+%
3-Methyl-1-butanol, ACS, 98.5+%
Thermo Scientific Chemicals

3-Methyl-1-butanol, ACS, 98.5+%

CAS: 123-51-3 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
Catalog number 036716.AP
also known as 036716-AP
Price (USD)
61.65
Online Exclusive
67.90
Save 6.25 (9%)
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
61.65
Online Exclusive
67.90
Save 6.25 (9%)
Each
Add to cart
Chemical Identifiers
CAS123-51-3
IUPAC Name3-methylbutan-1-ol
Molecular FormulaC5H12O
InChI KeyPHTQWCKDNZKARW-UHFFFAOYSA-N
SMILESCC(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Residual water≤0.5%
Impurity contentAcid and esters (as amyl acetate): ≤0.2%
Residue on Evaporation≤0.003%
Free acid (titration)≤0.002meq/g
View more
3-Methyl-1-butanol is used as an active ingredient in banana oil production and Kovac's reagent. It acts as an antifoaming agent. It serves as an intermediate for the manufacture of corrosion inhibitors, flotation aids, fuel and lubricating oil additives, polyvinyl chloride stabilizers, pharmaceuticals, cosmetics, plasticizers, rubber auxiliaries and paint solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only