Calcium phosphate tribasic, 34-40% Ca
Calcium phosphate tribasic, 34-40% Ca
Calcium phosphate tribasic, 34-40% Ca
Thermo Scientific Chemicals

Calcium phosphate tribasic, 34-40% Ca

CAS: 12167-74-7 | Ca3O8P2 | 310.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
10 kg
Catalog number 036731.A9
also known as 036731-A9
Price (USD)
627.00
Each
Quantity:
10 kg
Request bulk or custom format
Price (USD)
627.00
Each
Chemical Identifiers
CAS12167-74-7
IUPAC Nametricalcium diphosphate
Molecular FormulaCa3O8P2
InChI KeyQORWJWZARLRLPR-UHFFFAOYSA-H
SMILES[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)34-40% Ca
Utilized in powdered spices as an anticaking agent and baby powder. It is the starting material for the production of phosphoric acid and fertilizers. Employed in tissue replacement for repairing bony defects and act as drug carrier for drug delivery to the bone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only