Semiquantitative Standard 2, Specpure™
Thermo Scientific Chemicals

Semiquantitative Standard 2, Specpure™

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036770.AP
also known as 036770-AP
Price (USD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialSemiquantitative Standard 2
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, OXIDIZING, N.O.S.
Health Hazard 1Specific target organ toxicity after single exposure (category 3), Corrosive to metals (category 1), Skin corrosion/irritation (category 1)
Health Hazard 2May cause respiratory irritation, May be corrosive to metals, Causes severe skin burns and eye damage
MDL NumberMFCD02091743
View more
Semiquantitative Standard 2 is utilized as standard solution in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only