Mercury(II) thiocyanate
Mercury(II) thiocyanate
Mercury(II) thiocyanate
Thermo Scientific Chemicals

Mercury(II) thiocyanate

CAS: 592-85-8 | C4HgK2N4S4 | 511.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 037106.30
also known as 037106-30
Price (USD)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS592-85-8
IUPAC Namemercury(2+) dipotassium tetrakis(cyanosulfanide)
Molecular FormulaC4HgK2N4S4
InChI KeyHWXOCCNUUYRBKP-UHFFFAOYSA-J
SMILES[K+].[K+].[Hg++].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (special)61.7-64.9% Hg
FormPowder
Mercury(II) thiocyanate is used as a precursor to potassium tris(thiocyanato)mercurate(II) and cesium tris(thiocyanato)mercurate(II). It is also used in the determination of chloride ions in water by UV-visible spectroscopy. Further, it acts as a catalyst for the addition of thiocyanic acid to alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only