Phenylmercury(II) chloride, 96%, Hg 63.5%
Phenylmercury(II) chloride, 96%, Hg 63.5%
Phenylmercury(II) chloride, 96%, Hg 63.5%
Thermo Scientific Chemicals

Phenylmercury(II) chloride, 96%, Hg 63.5%

CAS: 100-56-1 | C6H5ClHg | 313.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 037113.09
also known as 037113-09
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Specifications
CAS100-56-1
Chemical Name or MaterialPhenylmercury(II) chloride
ColorWhite
Concentration or Composition (by Analyte or Components)Hg 63.5%
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: PHENYLMERCURIC COMPOUNDS, N.O.S.
View more
The reaction of phenylmercury (II) chloride with acetophenonethiosemicarbazone (Hatsc) in ethanol in 1: 1 mole ratio undergoes symmetrisation forming the products, HgCl2 (Hatse) 2 and Ph2Hg instead of the anticipated compound PhHgCl (Hatse). A new method for the determination of chloride ion is based on the formation of phenylmercury(II) chloride, its extraction into chloroform and reaction with sodium diethyldithiocarbamate to form phenylmercury(II) diethyldithiocarbamate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only