Dionex™ MMS Regenerant Concentrates
Actual product may vary
Thermo Scientific™

Dionex™ MMS Regenerant Concentrates

Increase productivity and provide reliable analyses with Thermo Scientific™ MMS Regenerant Concentrates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
0371644 x 200 mL
039602500 mL
Catalog number 037164
Price (MXN)
-
Volume (Metric):
4 x 200 mL

Increase productivity and provide reliable analyses with Thermo Scientific™ MMS Regenerant Concentrates.

  • Formulated with high-purity, reagent-grade chemicals and water (18 MΩ-cm); quality-control checked for ionic contamination, are used
  • All standards are NIST™ traceable

Includes:

A Certificate of Analysis is provided with every standard and chelation chromatography reagent

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Concentration20 X
For Use With (Application)HPLC
HazardsCorrosive
QuantityPack of 4
Standard ComponentsSulfuric Acid Aqueous Solution
Standard Concentration0.5 N
StorageRoom Temperature
Volume (Metric)4 x 200 mL
TypeRegenerant Concentrate
Unit SizePack of 4