Glassy carbon spherical powder, 0.4-12 micron, type 2
Thermo Scientific Chemicals

Glassy carbon spherical powder, 0.4-12 micron, type 2

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 038008.18
Price (USD)
200.65
Special offer
Online exclusive
Ends: 28-Dec-2026
236.00
Save 35.35
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
200.65
Special offer
Online exclusive
Ends: 28-Dec-2026
236.00
Save 35.35
Each
Specifications
Chemical Name or MaterialGlassy carbon spherical powder
EINECS Number231-153-3
MDL NumberMFCD00133992
PackagingGlass bottle
Recommended StorageAmbient temperatures
View more
In typical measurements, 8 cm downstream of the burner, of species profiles versus λ for a flame doped with type 1 glassy carbon spherical powder, as parts per million, normalized to the Ar36 peak area and the measured particle cross sectional area in Computer Model Study of Carbon Combustion in Low Pressure Laminar H 2+ O 2 Flames. Glassy carbon spherical powder (10-20 μm diameter) modified with cysteine methyl ester is found to be an inexpensive, novel material for the rapid removal of large quantities of toxic heavy metal ions such as Cd(II), Cu(II) and As(III) from aqueous media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only