Glassy carbon plate, 1mm (0.04in) thick, type 2
Glassy carbon plate, 1mm (0.04in) thick, type 2
Glassy carbon plate, 1mm (0.04in) thick, type 2
Thermo Scientific Chemicals

Glassy carbon plate, 1mm (0.04in) thick, type 2

Used in casting, smelting of metal, hard alloy, powder and glass. Glassy carbon plate is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices. It also used in solar energy applications. | CAS: 7440-44-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
Catalog number 038021.GH
also known as 038021-GH
Price (USD)
1,463.65
Online Exclusive
1,626.00
Save 162.35 (10%)
Each
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)
1,463.65
Online Exclusive
1,626.00
Save 162.35 (10%)
Each
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPlate, 1mm (0.04in) thick (Glassy Carbon type-2, maximimum service temperature 3000?C).
Appearance (Color)Black
Assay from Supplier's CofA>99.9%
Used in casting, smelting of metal, hard alloy, powder and glass. Glassy carbon plate is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices. It also used in solar energy applications. It offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the difference between type 1 glassy carbon and type 2 glassy carbon?

Type 1 is a "low temperature" glassy carbon produced by heat treatment at ~ 1,000-2,000°C and consists of discreet fragments of distorted graphene layers. In general, type 1 is less compressible, more resistant to oxidation, and less permeable to gasses. Type 2 is a"high temperature" glassy carbon produced by heat treatment at ~2,500-3,000°C and consists of broken or imperfect fullerene-like nanospheroids encased in a disordered, multilayered graphene matrix. Type 2 usually contains fewer impurities and has higher electrical and thermal conductivity.