Glassy carbon plate, 4mm (0.16in) thick, type 2
Thermo Scientific Chemicals

Glassy carbon plate, 4mm (0.16in) thick, type 2

Used as fuel cell and other electrochemical electrodes. It is also used in mechanical applications such as when used as a structural component of a furnace. It is also used as also used for certain thin film deposition experiments.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 038023.GH
also known as 038023-GH
Price (USD)
1,224.65
Online exclusive
1,360.00
Save 135.35
Each
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)
1,224.65
Online exclusive
1,360.00
Save 135.35
Each
Specifications
Chemical Name or MaterialGlassy carbon plate
EINECS Number231-153-3
MDL NumberMFCD00133992
PackagingTube
Recommended StorageAmbient temperatures
View more
Glassy carbon plate, is used as fuel cell and other electrochemical electrodes. It is also used in mechanical applications such as when used as a structural component of a furnace. It is also used as also used for certain thin film deposition experiments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the difference between type 1 glassy carbon and type 2 glassy carbon?

Type 1 is a "low temperature" glassy carbon produced by heat treatment at ~ 1,000-2,000°C and consists of discreet fragments of distorted graphene layers. In general, type 1 is less compressible, more resistant to oxidation, and less permeable to gasses. Type 2 is a"high temperature" glassy carbon produced by heat treatment at ~2,500-3,000°C and consists of broken or imperfect fullerene-like nanospheroids encased in a disordered, multilayered graphene matrix. Type 2 usually contains fewer impurities and has higher electrical and thermal conductivity.