Glassy carbon plate, 4mm (0.16in) thick, type 1
Glassy carbon plate, 4mm (0.16in) thick, type 1
Glassy carbon plate, 4mm (0.16in) thick, type 1
Thermo Scientific Chemicals

Glassy carbon plate, 4mm (0.16in) thick, type 1

CAS: 7440-44-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 038026.FI
also known as 038026-FI
Price (USD)
384.65
Online Exclusive
427.00
Save 42.35 (10%)
Each
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)
384.65
Online Exclusive
427.00
Save 42.35 (10%)
Each
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Total Metal Impurities≤0.1% (ash content)
FormPlate, 4mm (0.16in) thick (Glassy Carbon type-1, maximimum service temperature 1000°C).
Glassy carbon plate, is used as fuel cell and Due to their specific surface orientation, glassy carbon is employed as an electrode material for the fabrication of sensors. It is also used in mechanical applications such as when used as a structural component of a furnace. It is also used as also used for certain thin film deposition experiments. Chemically modified glasssy electrodes have been employed for the analysis of organic molecules (viz., Paracetamol, aspirin, caffeine, phenol, catechol, resorcinol, hydroquinone, dopamine, L-dopa, epinephrine, nor epinephrine, methyl parathion, ethyl parathion, venlafaxine, desvenlafaxine, imipramine, trimipramine, desipramine etc.) as well as metal ions (bismuth, antimony etc.)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the difference between type 1 glassy carbon and type 2 glassy carbon?

Type 1 is a "low temperature" glassy carbon produced by heat treatment at ~ 1,000-2,000°C and consists of discreet fragments of distorted graphene layers. In general, type 1 is less compressible, more resistant to oxidation, and less permeable to gasses. Type 2 is a"high temperature" glassy carbon produced by heat treatment at ~2,500-3,000°C and consists of broken or imperfect fullerene-like nanospheroids encased in a disordered, multilayered graphene matrix. Type 2 usually contains fewer impurities and has higher electrical and thermal conductivity.