Search
Search

| Catalog Number | Description |
|---|---|
| 038098.KT also known as 038098-KT | Watch glass; Fits 1000mL Beakers |
| 038099.KT also known as 038099-KT | Watch glass; Fits 2000mL Beakers |
| 038097.KT also known as 038097-KT | Watch glass; Fits 500mL Beakers |
| 038095.KT also known as 038095-KT | Watch glass; Fits 100mL Beakers |
| 038096.KT also known as 038096-KT | Watch glass; Fits 250mL Beakers |