Cerium(III) ammonium nitrate tetrahydrate, Reagent Grade
Cerium(III) ammonium nitrate tetrahydrate, Reagent Grade
Cerium(III) ammonium nitrate tetrahydrate, Reagent Grade
Thermo Scientific Chemicals

Cerium(III) ammonium nitrate tetrahydrate, Reagent Grade

CAS: 13083-04-0 | CeH16N7O19 | 558.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 038367.22
also known as 038367-22
Price (USD)
56.20
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
56.20
Each
Chemical Identifiers
CAS13083-04-0
IUPAC Namecerium(3+) bis(nitric acid) diamine tetrahydrate trinitrate
Molecular FormulaCeH16N7O19
InChI KeyAIVZRMAXALRCHB-UHFFFAOYSA-N
SMILESN.N.O.O.O.O.[Ce+3].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98% min
A useful oxidant for organic synthesis. This chemical is also useful for the synthesis of quinoxaline derivatives which are useful as DNA cleaving agents, dyes, and organic semiconductors. Used to release organic ligands from metal carbonyls. Whereby the metal is oxidized, Carbon monoxide is produced, and the organic ligand is released.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only