Platinum powder, -325 mesh, 99.9+% (metals basis)
Platinum powder, -325 mesh, 99.9+% (metals basis)
Platinum powder, -325 mesh, 99.9+% (metals basis)
Thermo Scientific Chemicals

Platinum powder, -325 mesh, 99.9+% (metals basis)

CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.1 g
1 g
5 g
Catalog number 038374.03
also known as 038374-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-325 mesh (less than 45 micron)
Trace MetalLess than 0.1% by wt trace metals
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only