Potassium chloride crystal optic disc, 25mm x 4mm, polished both sides
Thermo Scientific Chemicals

Potassium chloride crystal optic disc, 25mm x 4mm, polished both sides

 
Have Questions?
Catalog NumberQuantity
038420.KT1 Ea.
Catalog number 038420.KT
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.40
Save 12.75 (15%)
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.40
Save 12.75 (15%)
Each
Specifications
Chemical Name or MaterialPotassium chloride crystal optic disc
EINECS Number231-211-8
MDL NumberMFCD00011360
Name Note25mm x 4mm, Polished both sides
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only