Potassium bromide crystal optic disc, 25mm x 4mm, polished both sides
Thermo Scientific Chemicals

Potassium bromide crystal optic disc, 25mm x 4mm, polished both sides

 
Have Questions?
Catalog NumberQuantity
038425.KT
also known as 038425-KT
1 Ea.
Catalog number 038425.KT
also known as 038425-KT
Price (USD)
97.65
Online exclusive
108.00
Save 10.35
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
97.65
Online exclusive
108.00
Save 10.35
Each
Specifications
Chemical Name or MaterialPotassium bromide crystal optic disc
EINECS Number231-830-3
Health Hazard 1H301-H315-H319-H335: Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
MDL NumberMFCD00011358
Name NotePolished on both sides
View more
Potassium bromide crystal optic disc is used for infrared transmission windows in gas and liquid sample cells used with infrared and FTIR spectrophotometers and for beam splitters for spectrophotometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only