Chemical Name or MaterialPotassium bromide crystal optic disc
EINECS Number231-830-3
Name Note32mm x 3mm (drilled), Polished both sides
Recommended StorageAmbient temperatures
TSCAYes
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.