Sodium chloride crystal optic disc, 25mm x 2mm, polished both sides
Sodium chloride crystal optic disc, 25mm x 2mm, polished both sides
Sodium chloride crystal optic disc, 25mm x 2mm, polished both sides
Thermo Scientific Chemicals

Sodium chloride crystal optic disc, 25mm x 2mm, polished both sides

 
Have Questions?
Catalog NumberQuantity
038449.KT
also known as 038449-KT
1 Ea.
Catalog number 038449.KT
also known as 038449-KT
Price (USD)
80.20
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
80.20
Each
Specifications
Chemical Name or MaterialSodium chloride crystal optic disc
EINECS Number231-598-3
Name Note25mm x 2mm, Polished both sides
Recommended StorageAmbient temperatures
TSCAYes
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only