Potassium bromide crystal optic disc, 25mm x 2mm, polished both sides
Thermo Scientific Chemicals

Potassium bromide crystal optic disc, 25mm x 2mm, polished both sides

 
Have Questions?
Catalog NumberQuantity
038451.KT1 Ea.
Catalog number 038451.KT
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Specifications
Chemical Name or MaterialPotassium bromide crystal optic disc
EINECS Number231-830-3
MDL NumberMFCD00011358
Name Note25mm x 2mm, Polished both sides
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only