Sodium chloride crystal optic rectangle, 30mm x 15mm x 4mm (drilled), polished both sides
Sodium chloride crystal optic rectangle, 30mm x 15mm x 4mm (drilled), polished both sides
Sodium chloride crystal optic rectangle, 30mm x 15mm x 4mm (drilled), polished both sides
Thermo Scientific Chemicals

Sodium chloride crystal optic rectangle, 30mm x 15mm x 4mm (drilled), polished both sides

CAS: 7647-14-5 | ClNa | 58.44 g/mol
Have Questions?
Catalog NumberQuantity
038461.KT1 Ea.
Catalog number 038461.KT
Price (USD)
83.30
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
83.30
Each
Specifications
CAS7647-14-5
ChEBICHEBI:26710
Chemical Name or MaterialSodium chloride crystal optic rectangle
EINECS Number231-598-3
IUPAC Namesodium chloride
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only