Potassium bromide crystal optic rectangle, 38.5mm x 19.5mm x 4mm, polished both sides
Potassium bromide crystal optic rectangle, 38.5mm x 19.5mm x 4mm, polished both sides
Potassium bromide crystal optic rectangle, 38.5mm x 19.5mm x 4mm, polished both sides
Thermo Scientific Chemicals

Potassium bromide crystal optic rectangle, 38.5mm x 19.5mm x 4mm, polished both sides

CAS: 7758-02-3 | BrK | 119.00 g/mol
Have Questions?
Catalog NumberQuantity
038469.KT1 Ea.
Catalog number 038469.KT
Price (USD)
89.10
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
89.10
Each
Specifications
CAS2-3-7758
ChEBICHEBI:32030
Chemical Name or MaterialPotassium bromide crystal optic rectangle
EINECS Number231-830-3
IUPAC Namepotassium;bromide
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only