Aluminum oxide substrate, 10x10x0.5mm, polished one side, A plane
Aluminum oxide substrate, 10x10x0.5mm, polished one side, A plane
Aluminum oxide substrate, 10x10x0.5mm, polished one side, A plane
Thermo Scientific Chemicals

Aluminum oxide substrate, 10x10x0.5mm, polished one side, A plane

Al2O3
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 Ea.
5 Ea.
Catalog number 038498.KT
also known as 038498-KT
Price (USD)
77.65
Special offer
Online exclusive
Ends: 28-Dec-2026
91.00
Save 13.35
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 28-Dec-2026
91.00
Save 13.35
Each
Specifications
Chemical Name or MaterialAluminum Oxide Substrate, A Plane
EINECS Number215-691-6
MDL NumberMFCD00003424
Molecular FormulaAl2O3
Name NotePolished One Side
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only