Aluminum trifluoromethanesulfonate, Thermo Scientific Chemicals
Aluminum trifluoromethanesulfonate, Thermo Scientific Chemicals
Aluminum trifluoromethanesulfonate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum trifluoromethanesulfonate, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
038568.09
also known as 038568-09
10 g
Catalog number 038568.09
also known as 038568-09
Price (USD)
87.65
Online Exclusive
97.10
Save 9.45 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
87.65
Online Exclusive
97.10
Save 9.45 (10%)
Each
Specifications
CAS74974-61-1
Chemical Name or MaterialAluminum trifluoromethanesulfonate
ColorWhite
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Formula Weight474.2
View more
Aluminum trifluoromethanesulfonate is used in pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only