Bromocresol Green sodium salt, 0.04% w/v aq. soln.
Bromocresol Green sodium salt, 0.04% w/v aq. soln.
Bromocresol Green sodium salt, 0.04% w/v aq. soln.
Thermo Scientific Chemicals

Bromocresol Green sodium salt, 0.04% w/v aq. soln.

CAS: 62625-32-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 038696.AP
also known as 038696-AP
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
58.60
Save 8.95 (15%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
58.60
Save 8.95 (15%)
Each
Chemical Identifiers
CAS62625-32-5
IUPAC Namesodium 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-3-methylbenzen-1-olate
Molecular FormulaC21H13Br4NaO5S
InChI KeyHEFSGAHJDGZCHA-UHFFFAOYNA-M
SMILES[Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green to green-blue to blue
FormLiquid
Assay from Suppliers CofA0.040% w/v ± 0.002% w/v
CommentGreen/yellow at pH 3.8, Blue at pH 5.4
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only