Methyl Red, 0.1% w/v solution in ethanol
Methyl Red, 0.1% w/v solution in ethanol
Methyl Red, 0.1% w/v solution in ethanol
Thermo Scientific Chemicals

Methyl Red, 0.1% w/v solution in ethanol

CAS: 74886-84-3
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 038698.AE
also known as 038698-AE
Price (USD)
31.30
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
31.30
Each
Specifications
CAS74886-84-3
Chemical Name or MaterialMethyl Red
Concentration0.1% w/v solution in ethanol
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
Formula Weight269.3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use