Phenol Red sodium salt, 0.02% w/v aq. soln.
Phenol Red sodium salt, 0.02% w/v aq. soln.
Phenol Red sodium salt, 0.02% w/v aq. soln.
Thermo Scientific Chemicals

Phenol Red sodium salt, 0.02% w/v aq. soln.

CAS: 34487-61-1 | C19H13NaO5S | 376.358 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 038702.AP
also known as 038702-AP
Price (USD)
42.50
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
42.50
Each
Specifications
CAS34487-61-1
Chemical Name or MaterialPhenol Red sodium salt
Concentration0.02% w/v aq. soln.
Density0.972 g/mL
EINECS Number252-057-8
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only