Thymolphthalein, 0.05% w/v solution in ethanol
Thymolphthalein, 0.05% w/v solution in ethanol
Thymolphthalein, 0.05% w/v solution in ethanol
Thermo Scientific Chemicals

Thymolphthalein, 0.05% w/v solution in ethanol

CAS: 125-20-2 | C28H30O4 | 430.544 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 038706.AP
also known as 038706-AP
Price (USD)
61.65
Special offer
Online exclusive
Ends: 30-Jun-2026
72.10
Save 10.45 (14%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
61.65
Special offer
Online exclusive
Ends: 30-Jun-2026
72.10
Save 10.45 (14%)
Each
Chemical Identifiers
CAS125-20-2
IUPAC Name3,3-bis[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC28H30O4
InChI KeyLDKDGDIWEUUXSH-UHFFFAOYSA-N
SMILESCC(C)C1=CC(=C(C)C=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration (w/v)0.05 ± 0.0025% w/v in ethanol
FormLiquid
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use