Alizarin Yellow R sodium salt
Alizarin Yellow R sodium salt
Alizarin Yellow R sodium salt
Thermo Scientific Chemicals

Alizarin Yellow R sodium salt

CAS: 1718-34-9 | C13H8N3NaO5 | 309.213 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Catalog number 038707.09
also known as 038707-09
Price (USD)
66.65
Online Exclusive
74.30
Save 7.65 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
66.65
Online Exclusive
74.30
Save 7.65 (10%)
Each
Chemical Identifiers
CAS1718-34-9
IUPAC Namesodium (3Z)-3-[2-(4-nitrophenyl)hydrazin-1-ylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
Molecular FormulaC13H8N3NaO5
InChI KeyHKSANXZSKDUDBD-SOCRLDLMSA-M
SMILES[Na+].[O-]C(=O)C1=CC(C=CC1=O)=N/NC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color ChangepH 10.1 (yellow)
Color ChangepH 12.1 (orange)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only