Tropaeolin O
Tropaeolin O
Tropaeolin O
Thermo Scientific Chemicals

Tropaeolin O

CAS: 547-57-9 | C12H10N2NaO5S | 317.271 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 038708.04
also known as 038708-04
Price (USD)
24.00
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
24.00
Each
Chemical Identifiers
CAS547-57-9
IUPAC Name4-[2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]benzene-1-sulfonic acid sodium
Molecular FormulaC12H10N2NaO5S
InChI KeyYOGUPPQYRWYKGT-UHFFFAOYSA-N
SMILES[Na].OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only