Silicon, plasma standard solution, Specpure™ Si 1000μg/mL
Silicon, plasma standard solution, Specpure™ Si 1000μg/mL
Silicon, plasma standard solution, Specpure™ Si 1000μg/mL
Thermo Scientific Chemicals

Silicon, plasma standard solution, Specpure™ Si 1000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 038717.AP
also known as 038717-AP
Price (USD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialSilicon plasma standard solution
MDL NumberMFCD00085311
Name NoteSi 1000μg/mL
Physical FormLiquid
Quantity500 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only