Aluminum, plasma standard solution, Specpure™ Al 10,000μg/mL
Aluminum, plasma standard solution, Specpure™ Al 10,000μg/mL
Aluminum, plasma standard solution, Specpure™ Al 10,000μg/mL
Thermo Scientific Chemicals

Aluminum, plasma standard solution, Specpure™ Al 10,000μg/mL

Used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry. | CAS: 11145-27-0 | AlSi | 55.067 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 038721.AP
also known as 038721-AP
Price (USD)
246.65
Special offer
Online exclusive
Ends: 30-Jun-2026
290.00
Save 43.35 (15%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
246.65
Special offer
Online exclusive
Ends: 30-Jun-2026
290.00
Save 43.35 (15%)
Each
Specifications
CAS11145-27-0
Chemical Name or MaterialAluminum
DOT InformationHazard Class: 8
EINECS Number231-714-2
Health Hazard 1H290-H314-H335
View more
Aluminum, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only