Silicon, plasma standard solution, Specpure™ Si 10,000μg/mL
Silicon, plasma standard solution, Specpure™ Si 10,000μg/mL
Silicon, plasma standard solution, Specpure™ Si 10,000μg/mL
Thermo Scientific Chemicals

Silicon, plasma standard solution, Specpure™ Si 10,000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 038723.AD
also known as 038723-AD
Price (USD)
95.65
Special offer
Online exclusive
Ends: 30-Jun-2026
112.00
Save 16.35 (15%)
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
95.65
Special offer
Online exclusive
Ends: 30-Jun-2026
112.00
Save 16.35 (15%)
Each
Specifications
Chemical Name or MaterialSilicon plasma standard solution
Health Hazard 1H302
Health Hazard 3P264b-P270-P301+P312-P330-P501c
MDL NumberMFCD00085311
Name NoteSi 10000μg/mL
View more
Silicon plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only