Molybdenum, plasma standard solution, Specpure™ Mo 10,000μg/mL
Molybdenum, plasma standard solution, Specpure™ Mo 10,000μg/mL
Molybdenum, plasma standard solution, Specpure™ Mo 10,000μg/mL
Thermo Scientific Chemicals

Molybdenum, plasma standard solution, Specpure™ Mo 10,000μg/mL

CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 038726.AP
also known as 038726-AP
Price (USD)
244.65
Special offer
Online exclusive
Ends: 30-Jun-2026
288.00
Save 43.35 (15%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
244.65
Special offer
Online exclusive
Ends: 30-Jun-2026
288.00
Save 43.35 (15%)
Each
Specifications
CAS7439-98-7
ChEBICHEBI:28685
Chemical Name or MaterialMolybdenum
Health Hazard 1H315-H319
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only