Aluminum, plasma standard solution, Specpure™, Al 1000 mg/ml
Aluminum, plasma standard solution, Specpure™, Al 1000 mg/ml
Aluminum, plasma standard solution, Specpure™, Al 1000 mg/ml
Thermo Scientific Chemicals

Aluminum, plasma standard solution, Specpure™, Al 1000 mg/ml

CAS: 7784-27-2 | AlH18N3O18 | 375.129 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 038727.AD
also known as 038727-AD
Price (USD)
56.30
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
56.30
Each
Specifications
CAS7784-27-2
CAS7697-37-2
CAS7732-18-5
Chemical Name or MaterialAluminum
ColorColorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only