Sulfur in Diesel Fuel standard solution, Specpure™, 300μg/g (0.030%)
Thermo Scientific Chemicals

Sulfur in Diesel Fuel standard solution, Specpure™, 300μg/g (0.030%)

 
Have Questions?
Catalog NumberQuantity
038767.AE
also known as 038767-AE
100 mL
Catalog number 038767.AE
also known as 038767-AE
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Specifications
Chemical Name or MaterialSulfur in Diesel Fuel standard solution
EINECS Number270-676-1
Health Hazard 1H227-H351
Health Hazard 2GHS H Statement
H351-H315-H319-H335-H227
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P201-P202-P210-P281-P308+P313-P370+P378q-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only