Silicon rod, 2.5cm (0.98in) dia, 99.999% (metals basis)
Silicon rod, 2.5cm (0.98in) dia, 99.999% (metals basis)
Silicon rod, 2.5cm (0.98in) dia, 99.999% (metals basis)
Thermo Scientific Chemicals

Silicon rod, 2.5cm (0.98in) dia, 99.999% (metals basis)

CAS: 7440-21-3 | Si | 28.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mm
100 mm
150 mm
Catalog number 039102.DO
also known as 039102-DO
Price (USD)
686.65
Online Exclusive
763.00
Save 76.35 (10%)
Each
Quantity:
150 mm
Request bulk or custom format
Price (USD)
686.65
Online Exclusive
763.00
Save 76.35 (10%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-21-3
ChEBICHEBI:27573
Chemical Name or MaterialSilicon rod
EINECS Number231-130-8
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only