1,4-Dioxane, ACS, 99+%, stab. with 1-3ppm BHT
1,4-Dioxane, ACS, 99+%, stab. with 1-3ppm BHT
1,4-Dioxane, ACS, 99+%, stab. with 1-3ppm BHT
Thermo Scientific Chemicals

1,4-Dioxane, ACS, 99+%, stab. with 1-3ppm BHT

CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
4 x 500 mL
Catalog number 039118.K7
also known as 039118-K7
Price (USD)
242.65
Online Exclusive
269.00
Save 26.35 (10%)
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
242.65
Online Exclusive
269.00
Save 26.35 (10%)
Each
Chemical Identifiers
CAS123-91-1
IUPAC Name1,4-dioxane
Molecular FormulaC4H8O2
InChI KeyRYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILESC1COCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on Evaporation≤0.005%
Free acid (titration)≤0.0016 meq/g
Impurity contentCarbonyl (as HCHO): ≤0.01%
Impurity contentPeroxide: ≤0.005%
Water Content (Karl Fischer Titration)≤0.05%
View more
1,4-Dioxane is used as a stabilizer for 1,1,1-trichloroethane and chlorinated solvents. It acts as a solvent for cellulose acetate, ethyl cellulose, benzyl cellulose, resins, oils, waxes, dyes, other organic and inorganic compounds. It acts as a substitute for tetrahydrofuran due to its higher boiling point and lower toxicity. Furthermore, it is used as an internal standard for proton NMR spectroscopy in deuterium oxide. It plays an important role in liquid scintillation counting in biological procedures and as a wetting and dispersing agent in textile processing. It is involved in the preparation of dimethylmagnesium by reacting with methylmagnesium bromide and in the preparation of histological sections for microscopic examination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only