Ammonium copper(II) chloride dihydrate
Ammonium copper(II) chloride dihydrate
Ammonium copper(II) chloride dihydrate
Thermo Scientific Chemicals

Ammonium copper(II) chloride dihydrate

CAS: 10060-13-6 | Cl4CuH12N2O2 | 277.454 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 039127.36
also known as 039127-36
Price (USD)
329.65
Special offer
Online exclusive
Ends: 30-Jun-2026
388.00
Save 58.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
329.65
Special offer
Online exclusive
Ends: 30-Jun-2026
388.00
Save 58.35 (15%)
Each
Chemical Identifiers
CAS10060-13-6
IUPAC Namediammonium tetrachlorocopperdiuide dihydrate
Molecular FormulaCl4CuH12N2O2
InChI KeyLLMDTWKBBWCWNN-UHFFFAOYSA-L
SMILES[NH4+].[NH4+].O.O.Cl[Cu--](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)22.9%-23.7% Cu
Ammonium copper(II) chloride dihydrate is used in the determination iron of content in steel. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only