Lead(II) titanium(IV) isopropoxide, 99.9% (metals basis), 5% w/v in isopropanol
Thermo Scientific Chemicals

Lead(II) titanium(IV) isopropoxide, 99.9% (metals basis), 5% w/v in isopropanol

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number 039153.AE
also known as 039153-AE
Price (USD)
295.65
Special offer
Online exclusive
Ends: 30-Jun-2026
348.00
Save 52.35 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
295.65
Special offer
Online exclusive
Ends: 30-Jun-2026
348.00
Save 52.35 (15%)
Each
Specifications
Assay Percent Notes(metals basis)
Chemical Name or MaterialLead(II) titanium(IV) isopropoxide
ColorYellow to Brown
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
Flash Point12°C (54°F)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only