Molybdenum(V) isopropoxide, 99.6% (metals basis), 5% w/v in isopropanol
Molybdenum(V) isopropoxide, 99.6% (metals basis), 5% w/v in isopropanol
Molybdenum(V) isopropoxide, 99.6% (metals basis), 5% w/v in isopropanol
Thermo Scientific Chemicals

Molybdenum(V) isopropoxide, 99.6% (metals basis), 5% w/v in isopropanol

CAS: 209733-38-0 | C15H35MoO5 | 391.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number 039159.AE
also known as 039159-AE
Price (USD)
458.65
Online Exclusive
509.00
Save 50.35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
458.65
Online Exclusive
509.00
Save 50.35 (10%)
Each
Chemical Identifiers
CAS209733-38-0
IUPAC Namepentakis(propan-2-yloxy)molybdenum
Molecular FormulaC15H35MoO5
InChI KeyHLFWWCCBFJUYJL-UHFFFAOYSA-N
SMILESCC(C)O[Mo](OC(C)C)(OC(C)C)(OC(C)C)OC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to orange to brown
FormLiquid
Assay from Suppliers CofA5% ± 0.5% [Molybdenum (V) iso-propoxide]
Total Metal Impurities≤0.4% (≤4000ppm)
Molybdenum(V) isopropoxide is used to prepare novel MoVSbOxtype catalysts for selective isobutane oxidation. It is used to prepare molybdenum Oxide Nanofibers by electrospinning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only