Tin(IV) 2-ethylhexanoate diisopropoxide, 98% (metals basis)
Thermo Scientific Chemicals

Tin(IV) 2-ethylhexanoate diisopropoxide, 98% (metals basis)

CAS: 288374-32-3
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 039169.06
also known as 039169-06
Price (USD)
103.65
Special offer
Online exclusive
Ends: 30-Jun-2026
122.00
Save 18.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
103.65
Special offer
Online exclusive
Ends: 30-Jun-2026
122.00
Save 18.35 (15%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS288374-32-3
Chemical Name or MaterialTin(IV) 2-ethylhexanoate diisopropoxide
Formula Weight523.08
Health Hazard 1H315-H319-H361d
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only