1-Methyl-2-pyrrolidinone, Spectrophotometric Grade, 99.5+%
1-Methyl-2-pyrrolidinone, Spectrophotometric Grade, 99.5+%
1-Methyl-2-pyrrolidinone, Spectrophotometric Grade, 99.5+%
1-Methyl-2-pyrrolidinone, Spectrophotometric Grade, 99.5+%
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, Spectrophotometric Grade, 99.5+%

For Spectrophotometry/ Chromatography | CAS: 872-50-4 | C5H9NO | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039176.K2
also known as 039176-K2
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS872-50-4
IUPAC Name1-methylpyrrolidin-2-one
Molecular FormulaC5H9NO
InChI KeySECXISVLQFMRJM-UHFFFAOYSA-N
SMILESCN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water0.05%
Residue on Evaporation6ppm
1-methyl-2-pyrrolidinone, is useful for spectrophotometry, chromatography and ICP-MS detection. It is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Evaporation residue 6ppm, H2O 0.05%
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only