Silver foil, 0.5mm (0.02in) thick, annealed, 99.9% (metals basis)
Silver foil, 0.5mm (0.02in) thick, annealed, 99.9% (metals basis)
Silver foil, 0.5mm (0.02in) thick, annealed, 99.9% (metals basis)
Thermo Scientific Chemicals

Silver foil, 0.5mm (0.02in) thick, annealed, 99.9% (metals basis)

Form: Approx. 3.3g/25 x 25mm; Temper: soft | CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
200 x 200 mm
Catalog number 039181.FF
also known as 039181-FF
Price (USD)
-
Quantity:
25 x 25 mm
Request bulk or custom format
Specifications
AppearanceSilver-colored foil
Assay Percent Range99.9% (Metals basis)
CAS7440-22-4
ChEBICHEBI:9141
Chemical Name or MaterialSilver
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only