Cesium tungsten oxide, 99.9% (metals basis)
Cesium tungsten oxide, 99.9% (metals basis)
Cesium tungsten oxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Cesium tungsten oxide, 99.9% (metals basis)

CAS: 13587-19-4 | Cs2O4W
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 039206.06
also known as 039206-06
Price (USD)
83.65
Special offer
Online exclusive
Ends: 28-Dec-2026
98.70
Save 15.05
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
83.65
Special offer
Online exclusive
Ends: 28-Dec-2026
98.70
Save 15.05
Each
Chemical Identifiers
CAS13587-19-4
IUPAC Namedicaesium(1+) tungsten tetraoxidandiide
Molecular FormulaCs2O4W
InChI KeyZYAJNDITUUBDPX-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[Cs+].[Cs+].[W]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Cesium tungsten oxide is used for making transparent insulation coating that has the best infrared transparent barrier feature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only