Bismuth vanadium oxide, 99.9% (metals basis)
Bismuth vanadium oxide, 99.9% (metals basis)
Bismuth vanadium oxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Bismuth vanadium oxide, 99.9% (metals basis)

CAS: 14059-33-7 | BiO4V | 323.918 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 039220.22
also known as 039220-22
Price (USD)
353.65
Online Exclusive
393.00
Save 39.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
353.65
Online Exclusive
393.00
Save 39.35 (10%)
Each
Chemical Identifiers
CAS14059-33-7
IUPAC Namebismuth(3+) vanadium tetraoxidandiide
Molecular FormulaBiO4V
InChI KeyHUUOUJVWIOKBMD-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[V].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Bismuth vanadium oxide is used as a pigment due to its hiding power. It also finds use in paints, coatings, plastic and rubber products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only