Bis(1,5-cyclooctadiene)nickel(0)
Bis(1,5-cyclooctadiene)nickel(0)
Bis(1,5-cyclooctadiene)nickel(0)
Bis(1,5-cyclooctadiene)nickel(0)
Thermo Scientific Chemicals

Bis(1,5-cyclooctadiene)nickel(0)

CAS: 1295-35-8 | C16H24Ni | 275.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 039234.04
also known as 039234-04
Price (USD)
120.00
Each
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)
120.00
Each
Add to cart
Chemical Identifiers
CAS1295-35-8
IUPAC Namebis((1Z,5Z)-cycloocta-1,5-diene) nickel
Molecular FormulaC16H24Ni
InChI KeyJRTIUDXYIUKIIE-KZUMESAESA-N
SMILES[Ni].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
View more
SpecificationsSpecification SheetSpecification Sheet
Gravimetric analysisNi=20.9-21.8 wt%
Appearance (Color)Yellow to orange.
FormCrystalline powder.
Gravimetric analysis>98.0%
Bis(1,5-cyclooctadiene)nickel(0) is used as a catalyst for the cycloaddition of 1,3-dienes and is used to catalyze the addition of allyl phenyl sulfide to alkynes leading to 1,4-dienes. Also, it is involved as a catalyst for asymmetric alpha-arylation and heteroarylation of ketones with chloroarenes, stereoselective borylative ketone-diene coupling, cycloaddition of benzamides with internal alkynes and methyl carboxylation of homopropargylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only