Bis(triphenylphosphine)dicarbonylnickel, 98%
Bis(triphenylphosphine)dicarbonylnickel, 98%
Bis(triphenylphosphine)dicarbonylnickel, 98%
Thermo Scientific Chemicals

Bis(triphenylphosphine)dicarbonylnickel, 98%

CAS: 13007-90-4 | C38H30NiO2P2 | 639.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
50 g
Catalog number 039242.09
also known as 039242-09
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
107.00
Save 16.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
107.00
Save 16.35 (15%)
Each
Specifications
CAS13007-90-4
Chemical Name or MaterialBis(triphenylphosphine)dicarbonylnickel
DOT InformationTransport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
EINECS Number235-851-9
Formula Weight639.29
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only